Learn More
HSP70 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532446
Code : PN
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast carcinoma.
HSP70 is a Heat shock protein (HSP) which are expressed in response to various biological stresses, including high temperatures. There are several major families of HSPs that include HSP70, there are HSP90 and HSP100. The HSP70 family is a set of highly conserved proteins that are induced by a variety of biological stresses, including heat stress, in every organism in which the proteins have been examined. The human HSP70 family members include: HSP70, a protein which is strongly inducible in all organisms but which is also constitutively expressed in primate cells; HSP72, a 72 kDa protein that is induced exclusively under stress conditions; HSC70, or cognate protein, is a 72 kDa, constitutively expressed, protein which is involved in the uncoating of clathrin coated vesicles; GRP78, or BiP, is a glucose regulated 78 kDa protein localized in the endoplasmic reticulum; and p75, or HSP75, a 75 kDa protein that is found within the mitochondria. Further, HSP70 is encoded by an intronless gene and, in conjunction with other heat shock proteins, HSP70 stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and organelles. HSP70 is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The HSP70 gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.Specifications
HSP70 | |
Polyclonal | |
HSPA1A | |
HSP70-1; HSP70-1A; HSP70I; HSP72; HSPA1; HSP70-1/HSP70-2; HSP70.1/HSP70.2; dnaK-type molecular chaperone HSP70-1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A/1B; heat shock 70kD protein 1A; heat shock-induced protein, Hspa70 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3303, 24472, 193740 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
P0DMV8, P0DMW0, Q61696 | |
HSPA1A | |
Synthetic peptide corresponding to aa 574-591 of human HSP70 | |
500 μL | |
Primary | |
Canine, Human, Mouse, Non-human Primate | |
Antibody | |
IgG |