Learn More
Armenian Hamster IgG, PE, Isotype Control, Invitrogen™
Armenian Hamster Isotype Control
Brand: Invitrogen PA533222
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
PA5-33222 is suitable for use as a negative control for the measurement of non-specific binding of Armenian hamster monoclonal antibodies, of isotype IgG, in human, mouse or rat tissues.Reconstitute with 1 ml of distilled water. After reconstitution, store product undiluted at 4ºC in the dark.Specifications
Armenian Hamster IgG | |
0.1mg/mL | |
IgG | |
Trinitrophenol - KLH | |
100 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |
Flow Cytometry | |
PE | |
Armenian Hamster | |
Ion-exchange chromatography | |
RUO | |
None | |
Lyophilized |