missing translation for 'onlineSavingsMsg'
Learn More

HAI-2 Rat anti-Mouse, Clone: 170317, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1107

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448345

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HAI-2 Monoclonal antibody MAB1107 Specifically detects HAI-2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

HAI-2/SPINT2
170317
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
HAI2, HAI2A, HAI2B, HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2, SPINT2
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rmHAI-2 in direct ELISAs and Western blots. Less than 5% cross-reactivity was observed with rmHAI-1 and rhHAI-2.
Mouse
Western Blot
LYOPH
Western Blot 1 ug/mL
Purified
SPINT2
Mouse myeloma cell line NS0-derived recombinant mouse HAI-2B aa 30-197 (del 37-93)
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10653
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only