Learn More
HAI-2 Mouse anti-Human, Clone: 170018, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1106
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438345
  • £390.72 / 500µg
Estimated Shipment
13-08-2020
Description
Description
HAI-2 Monoclonal antibody MAB1106 Specifically detects HAI-2 in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
HAI-2/SPINT2
170018
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SPINT2
Mouse myeloma cell line NS0-derived recombinant human HAI-2 aa 28-197 Accession # O43291
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10653
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, ELISA
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O43291
HAI2, HAI2A, HAI2B, HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2, SPINT2
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects recombinant human (rh)HAI-2 in direct ELISAs and Western blots. In these formats, does not cross-react with rhHAI-1 or rmHAI-2B.