missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EHMT2 Monoclonal Antibody (A8620A), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 435200
Code : 50
Additional Details : Weight : 0.00010kg
Description
This antibody specifically recognizes mouse G9a and cross reacts with human G9a.
EHMT2 Monoclonal antibody specifically detects EHMT2 in Human, Mouse samples. It is validated for ChIP Assay, ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
EHMT2 | |
Monoclonal | |
1 mg/mL | |
physiological saline with 0.1% sodium azide | |
Q96KQ7, Q9Z148 | |
EHMT2 | |
Produced in BALB/c mouse ascites after inoculation with hybridoma of mouse myeloma cells (NS-1) and spleen cells derived from a BALB/c mouse immunized with Baculovirus-expressed recombinant mouse G9a (15-114aa). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
ChIP Assay, ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
A8620A | |
Unconjugated | |
EHMT2 | |
ankyrin repeat-containing protein; Bat8; C6orf30; D17Ertd710e; DKFZp686H08213; Ehmt2; euchromatic histone lysine methyltransferase 2; euchromatic histone lysine N-methyltransferase 2; euchromatic histone-lysine N-methyltransferase 2; FLJ35547; G9a; G9A histone methyltransferase; GAT8; H3 ly; H3-K9-Histone-Methyl-Transferase; H3-K9-HMTase 3; histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B associated transcript 8, rat orthologue; HLA-B-associated transcript 8; KMT1C; lysine N-methyltransferase 1C; NG36; NG36/G9a; Protein G9a | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
10919, 110147 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |