missing translation for 'onlineSavingsMsg'
Learn More

GTF2I, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA535200

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15337614

  • £304.30 / 400µL


Estimated Shipment
08-06-2018
Add to basket

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications

Specifications

GTF2I
Polyclonal
PBS with 0.09% sodium azide
P78347
GTF2I, BAP135, WBSCR6, General transcription factor II-I, Bruton tyrosine kinase-associated protein 135, SRF-Phox1-interacting protein, Williams-Beuren syndrome chromosomal region 6 protein
Rabbit
IgG
400μL
Antibody
Polyclonal
Human
ImmunocytochemistryþImmunofluorescenceþWestern Blot
Unconjugated
GTF2I
Purified
GTF2I
KLH conjugated synthetic peptide between 281-310 amino acids from the N-terminal region of human GTF2I
Antigen affinity chromatography
Store at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
2969
Videos