missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2I Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535321
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat based on sequence homology.
GTF2I Polyclonal antibody specifically detects GTF2I in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GTF2I | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P78347 | |
GTF2I | |
KLH conjugated synthetic peptide between 956-985 amino acids from the C-terminal region of human GTF2I | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
GTF2I | |
6030441I21Rik; BAP135; BAP-135; Bruton tyrosine kinase-associated protein 135; BTKAP1; BTK-associated protein 135; BTK-associated protein, 135kD; DIWS; Diws1t; general transcription factor II I; general transcription factor IIi; general transcription factor II-I; Gtf2i; Gtf2ird1; GTFII-I; IB291; OTTHUMP00000209509; Spin; SRF-Phox1-interacting protein; tfiii; TFII-I; WBS; WBSCR6; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2969 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |