missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534698
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T. Predicted reactivity: Dog (92%), Pig (93%), Rhesus Monkey (97%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GSTA4 Polyclonal antibody specifically detects GSTA4 in Human, Porcine samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
GSTA4 | |
Polyclonal | |
Unconjugated | |
GSTA4 | |
glutathione S-alkyltransferase A4; glutathione S-aralkyltransferase A4; glutathione S-aryltransferase A4; glutathione S-transferase 5.7; Glutathione S-transferase A4; glutathione S-transferase A4-4; glutathione S-transferase alpha 4; glutathione S-transferase, alpha 4; glutathione transferase A4-4; GST 5.7; GST A4-4; GST class-alpha member 4; GST5.7; Gsta; GSTA4; GSTA4-4; GTA4; mGsta4; S-(hydroxyalkyl)glutathione lyase A4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100152951, 2941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O15217 | |
GSTA4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 222 of GSTA4 (Uniprot ID#O15217) | |
100 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG |