missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK3 alpha Monoclonal Antibody (9D5G1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515338
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15338 targets GSK3 alpha in indirect ELISA, ICC and WB applications and shows reactivity with Human samples. The MA5-15338 immunogen is purified recombinant fragment of GSK3 alpha expressed in E. Coli.
GSK3 alpha Monoclonal antibody specifically detects GSK3 alpha in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
GSK3 alpha | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P49840 | |
Gsk3a | |
Purified recombinant fragment of GSK3 alpha expressed in E. Coli. | |
RUO | |
2931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
9D5G1 | |
Unconjugated | |
Gsk3a | |
2700086H06Rik; DKFZp686D0638; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 alpha b; glycogen synthase kinase 3 alpha b; glycogen synthase kinase-3 alpha; glycogen synthase kinase-3 alpha; GSK3; GSK-3 alpha; GSK-3[a]; GSK-3[b]; gsk3a; gsk3ab; gsk-3-alpha; OTTHUMP00000198368; Serine/threonine-protein kinase GSK3A | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |