missing translation for 'onlineSavingsMsg'
Learn More

Growth Hormone Mouse anti-Rat, Clone: 222517, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB15661

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488735

  • £353.77 / 500µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Growth Hormone Monoclonal antibody MAB15661 Specifically detects Growth Hormone in Rat samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Growth Hormone
222517
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
GH1
E. coli-derived recombinant rat Growth Hormone, Phe27-Phe216, Accession # P01244
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2688
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
P01244
GH1, GHIGHD1B, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, Somatotropin
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects rat Growth Hormone in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Growth Hormone is observed.
Rat
Videos

For Research Use Only