Learn More
Gram Positive Bacteria LTA Monoclonal Antibody (G43J), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17402
Code : LD
Additional Details : Weight : 1.24740kg
Description
The MA1-7402 antibody reacts with Gram positive bacteria LTA in bacterial and infected tissue samples. MA1-7402 has been successfully used in ELISA, Western blot, and immunofluorescence applications. Suggested, but not tested in immunohistochemistry applications. The MA1-7402 antibody was raised against Lipoteichoic acid consistent with that of gram positive bacteria.
Gram Positive Bacteria LTA Monoclonal antibody specifically detects Gram Positive Bacteria LTA in Bacteria samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Gram Positive Bacteria LTA | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Western Blot | |
G43J | |
Unconjugated | |
Lipoteichoic acid | |
Lipoteichoic acid consistent with that of gram pos. bacteria. | |
100 μg | |
Primary | |
4° C | |
Liquid |