Learn More
GPT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543270
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PEYSSNVELA SFHSTSKGYM GECGYRGGYM EVINLHPEIK GQLVKLLSVR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%
GPT2 Polyclonal antibody specifically detects GPT2 in Human samples. It is validated for Western Blot.Specifications
GPT2 | |
Polyclonal | |
Unconjugated | |
GPT2 | |
4631422C05Rik; AAT2; Alanine aminotransferase 2; ALT2; AU014768; AU041193; C87201; Cc2-5; glutamate pyruvate transaminase 2; glutamic pyruvate transaminase (alanine aminotransferase) 2; glutamic pyruvate transaminase 2; glutamic--alanine transaminase 2; glutamic-pyruvate transaminase 2; Glutamic--pyruvic transaminase 2; GPT 2; GPT2; im:7150662; LOW QUALITY PROTEIN: alanine aminotransferase 2; MRT49; sb:cb580; zgc:165657 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84706 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TD30 | |
GPT2 | |
Synthetic peptide directed towards the C-terminal of human GPT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |