missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR82 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533767
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey (93%) Marmoset, Elephant, Panda, Dog, Bovine, Rabbit, Pig (87%) Horse (80%).
GPR82 Polyclonal antibody specifically detects GPR82 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
GPR82 | |
Polyclonal | |
Unconjugated | |
GPR82 | |
G protein-coupled receptor 82; GPR82; Probable G-protein coupled receptor 82 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27197 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96P67 | |
GPR82 | |
Synthetic 15 amino acid peptide from 2nd cytoplasmic domain of human GPR82. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |