Learn More
GPR77 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532839
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human (100%); Gorilla (95%); Monkey (90%); Marmoset (85%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
GPR77 Polyclonal antibody specifically detects GPR77 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Specifications
GPR77 | |
Polyclonal | |
Unconjugated | |
C5AR2 | |
C5a anaphylatoxin chemotactic receptor 2; C5a anaphylatoxin chemotactic receptor C5L2; C5AR2; C5L2; Complement component 5a receptor 2; G protein-coupled receptor 77; G protein-coupled receptor C5L2; GPF77; GPR77; G-protein coupled receptor 77 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27202 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9P296 | |
C5AR2 | |
Synthetic 20 amino acid peptide from C-terminal cytoplasmic domain of human C5L2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |