missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR64 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533751
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster, Elephant, Opossum (100%) Bovine, Panda, Rabbit (95%).
GPR64 Polyclonal antibody specifically detects GPR64 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
GPR64 | |
Polyclonal | |
Unconjugated | |
ADGRG2 | |
ADGRG2; adhesion G protein-coupled receptor G2; adhesion G-protein coupled receptor G2; AW212196; B830041D06Rik; CTD-2245E12.1; EDDM6; epididymal protein 6; epididymis-specific protein 6; FLJ00282; G protein-coupled receptor 64; G protein-coupled receptor, epididymis-specific (seven transmembrane family); Gpr64; G-protein coupled receptor 64; He6; Human epididymis-specific protein 6; LNB-TM7 heptahelical receptor Me6; Me6; MGC104454; MGC138738; MGC138739; Mouse epididymis-specific protein 6; Rat epididymis-specific protein 6; Re6; re6 receptor; TM7LN2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10149, 237175, 266735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8CJ11, Q8CJ12, Q8IZP9 | |
ADGRG2 | |
Synthetic 20 amino acid peptide from C-terminus of human GPR64. | |
50 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |