missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR43 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533718
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%).
GPR43 Polyclonal antibody specifically detects GPR43 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
GPR43 | |
Polyclonal | |
Unconjugated | |
FFAR2 | |
fatty acid receptor 2; FFA2; FFA2R; FFAR2; free fatty acid activated receptor 2; free fatty acid receptor 2; G protein-coupled receptor 43; GPCR43; Gpr43; G-protein coupled receptor 43; leukocyte-specific STAT-induced GPCR; Lssig | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2867 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O15552 | |
FFAR2 | |
Synthetic 17 amino acid peptide from 3rd extracellular domain of human GPR43. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |