missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3003
Code : NEW
Additional Details : Weight : 0.00900kg
Description
IHC (P) analysis shows positive staining of GPR3 in human pancreatic duct tissue.
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR3 is an orphan receptor with constitutive G signaling activity. GPR3 modulates a number of brain functions, including behavioral responses to stress, neurite outgrowth and amyloid-beta peptide generation suggesting that it may play a role in Alzheimer′s disease.Specifications
GPR3 | |
Polyclonal | |
whole serum with 0.05% sodium azide | |
P46089 | |
GPR3 | |
KLH conjugated Cys-SSSKIPFRSRSPSDV | |
RUO | |
2827 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
GPR3 | |
GPR3, G protein-coupled receptor 3, G-protein coupled receptor 3, ACCA orphan receptor, ACCA, Adenylate Cyclase Constitutive Activator | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |