missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPAR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533895
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Bat, Hamster (100%).
LPAR4 Polyclonal antibody specifically detects LPAR4 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin).Specifications
LPAR4 | |
Polyclonal | |
Unconjugated | |
Lpar4 | |
5730485F04Rik; G protein-coupled receptor 23; Gpr23; G-protein coupled receptor 23; LPA receptor 4; LPA4; LPA-4; LPA4 Receptor; Lpar4; Lysophosphatidic acid receptor 4; P2RY9; P2Y purinoceptor 9; P2Y5-LIKE; P2Y5-like receptor; p2y9; Purinergic receptor 9; RGD1563686 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2846, 302378, 78134 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8BLG2, Q99677 | |
Lpar4 | |
Synthetic 19 amino acid peptide from N-terminal extracellular domain of human LPAR4 / GPR23. | |
50 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |