missing translation for 'onlineSavingsMsg'
Learn More

GPR149, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA533640

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15344711

  • £294.00 / 50µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bat, Pig (100%) Rabbit (93%) Elephant, Bovine (87%).
Specifications

Specifications

GPR149
Polyclonal
Unconjugated
GPR149
Purified
GPR149
Synthetic 15 amino acid peptide from N-terminal extracellular domain of human GPR149.
Antigen affinity chromatography
Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
344758
Immunohistochemistry (Paraffin)
1mg/mL
PBS with <0.1% sodium azide
Q86SP6
GPR149, PGR10, G protein-coupled receptor 149, IEDA
Rabbit
IgG
50μg
Antibody
Polyclonal
Human
Videos