missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA3004
Code : NEW
Additional Details : Weight : 0.00900kg
Description
IHC (P) analysis shows positive staining of GPR12 in human kidney tissue.
GPR12 Polyclonal antibody specifically detects GPR12 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
GPR12 | |
Polyclonal | |
Unconjugated | |
GPR12 | |
FLJ18149; FLJ97704; G protein-coupled receptor 12; GPCR01; GPCR12; Gpcr20; Gpcr21; Gpr12; G-protein coupled receptor 12; MGC138349; MGC138351; PPP1R84; protein phosphatase 1, regulatory subunit 84; R334 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P47775 | |
GPR12 | |
KLH conjugated Cys-IPSSLAQRARSPSDV | |
RUO | |
2835 | |
-20°C | |
Liquid |