Learn More
Gng12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529207
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (97%), Rat (95%), Xenopus laevis (81%), Pig (95%), Rhesus Monkey (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Gng12 Polyclonal antibody specifically detects Gng12 in Human, Mouse samples. It is validated for Western Blot.Specifications
Gng12 | |
Polyclonal | |
Unconjugated | |
GNG12 | |
2010305F15Rik; AA536815; AI115529; AI314170; AI842738; G protein subunit gamma 12; GNG12; G-protein gamma-12 subunit; guanine nucleotide binding protein (G protein), gamma 12; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14701, 55970 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9DAS9, Q9UBI6 | |
GNG12 | |
Full length human GNG12 Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |