Learn More
GNAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528336
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, mouse eye. Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (92%), Xenopus laevis (92%), Dog (99%), Chicken (96%), Bovine (98%), Guinea pig (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GNAT1 Polyclonal antibody specifically detects GNAT1 in Human, Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
GNAT1 | |
Polyclonal | |
Unconjugated | |
GNAT1 | |
CSNB1G; CSNBAD3; G protein subunit alpha transducin 1; GBT1; Gnat1; Gnat-1; GNATR; guanine nucleotide binding protein (G protein), alpha transducing 1; guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 1; guanine nucleotide binding protein, alpha transducing 1; guanine nucleotide-binding protein G(t) subunit alpha-1; guanine nucleotide-binding protein G(T), alpha-1 subunit; Ird1; Ird2; irdc; irdr; rod-type transducin alpha subunit; Tr alpha; Tralpha; transducin; transducin alpha subunit; transducin alpha-1 chain; transducin alpha-subunit; transducin, rod-specific; transducin-alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14685, 2779 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
1.34 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P11488, P20612 | |
GNAT1 | |
Recombinant fragment corresponding to a region within amino acids 37 and 350 of Human GNAT1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |