missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNA15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523985
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rabbit based on sequence homology.
GNA15 Polyclonal antibody specifically detects GNA15 in Human, Mouse samples. It is validated for Western Blot.Specifications
GNA15 | |
Polyclonal | |
Unconjugated | |
GNA15 | |
Epididymis tissue protein Li 17E; G alpha 15; G alpha 16; g alpha-15; G alpha-16; G protein subunit alpha 15; G[a]15; Galpha15; GNA 15; GNA 16; GNA15; Gna-15; GNA16; G-protein subunit alpha-15; G-protein subunit alpha-16; Gq class; guanine nucleotide binding protein (G protein), alpha 15 (Gq class); guanine nucleotide binding protein, alpha 15; guanine nucleotide-binding protein subunit alpha-15; Guanine nucleotide-binding protein subunit alpha-16 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
14676, 2769 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P30678, P30679 | |
GNA15 | |
KLH conjugated synthetic peptide between 333-361 amino acids from the C-terminal region of human GNA15 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |