Learn More
GMEB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530402
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (95%), Rat (96%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GMEB1 Polyclonal antibody specifically detects GMEB1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GMEB1 | |
Polyclonal | |
Unconjugated | |
GMEB1 | |
1110050A04Rik; AI256615; AI481278; DNA-binding protein p96PIF; glucocorticoid modulatory element binding protein 1; glucocorticoid modulatory element-binding protein 1; Gmeb1; GMEB-1; P96PIF; Parvovirus initiation factor p96; PIF p96; PIF96; RGD1563208 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10691, 56809 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9JL60, Q9Y692 | |
GMEB1 | |
Recombinant fragment corresponding to a region within amino acids 315 and 573 of Human GMEB1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |