missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525737
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with non-human primate based on sequence homology.
GM2A Polyclonal antibody specifically detects GM2A in Human, Mouse samples. It is validated for Western Blot.Specifications
GM2A | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P17900, Q60648 | |
GM2A | |
KLH conjugated synthetic peptide between 25-54 amino acids from the N-terminal region of human GM2A | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GM2A | |
AA408702; AW215435; cerebroside sulfate activator protein; Ganglioside GM2 activator; Ganglioside GM2 activator isoform short; GM2 ganglioside activator; GM2 ganglioside activator protein; Gm2a; GM2-AP; SAP-3; shingolipid activator protein 3; sphingolipid activator protein 3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
14667, 2760 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |