Learn More
GM-CSF Mouse anti-Canine, Clone: 249201, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1546
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13428725
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
GM-CSF Monoclonal antibody MAB1546 Specifically detects GM-CSF in Canine samples. It is validated for ELISA Capture
Specifications
Specifications
GM-CSF
249201
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CSF2
E. coli-derived recombinant canine GM-CSF Ala18-Lys144 Accession # P48749.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1437
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P48749.1
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim,
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects canine GM-CSF in ELISAs. In ELISAs, this antibody does not cross-react with recombinant human GM-CSF, recombinant mouse GM-CSF, recombinant rat GM-CSF, recombinant feline GM-CSF, recombinant porcine GM-CSF, or recombinant canine IL-5.
Canine