missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPX1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518532
Code : LD
Additional Details : Weight : 0.01000kg
Description
GPX1 Polyclonal antibody specifically detects GPX1 in Human, Porcine samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
GPX1 | |
Polyclonal | |
Unconjugated | |
GPX1 | |
AI195024; AL033363; Cellular glutathione peroxidase; CGPx; cytosolic glutathione peroxidase; glutathione peroxidase; glutathione peroxidase 1; Gpx; Gpx1; GPx-1; GPXD; GSHPx; GSHPX1; GSHPx-1; HGNC:4553; MGC14399; MGC88245; OTTHUMP00000210766; Selenium-dependent glutathione peroxidase 1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2876, 397403 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P07203, Q8MJ14 | |
GPX1 | |
Synthetic peptide sequence (REALPAPSDDATA) corresponding to the internal amino acids of GPX1 | |
100 μg | |
Primary | |
Human, Porcine | |
Antibody | |
IgG |