missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA124637
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-24637 is expected to cross react with human (100% conserved) and chicken (90% conserved) due to sequence homology.
mGluR5 Polyclonal antibody specifically detects mGluR5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
mGluR5 | |
Polyclonal | |
tris glycine with 1% BSA and 0.1% sodium azide | |
P31424, P41594, Q3UVX5 | |
Grm5 | |
Synthetic peptide corresponding to residues Y D R R L A Q H K S E I E of mGluR5. | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Grm5 | |
6430542K11Rik; A830007B11Rik; AI850523; D16Ium24; D16Ium24e; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member E; Glu5R; Glur5; Glur-5; Glurbeta1; glutamate metabotropic receptor 5; glutamate receptor; glutamate receptor, metabotropic 5; GPRC1A; GPRC1E; GRM 1; GRM 5; GRM1; GRM1 alpha; GRM1A; Grm5; metabotropic 1; metabotropic glutamate receptor (mGluR5); metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; MGC25101; mGlu1; mGlu5; mGluR; mGluR1; mGluR1 alpha; mGluR1A; MGLUR5; mGluR5a; mGluR5b; PPP1R86; protein phosphatase 1, regulatory subunit 86 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
108071, 24418, 2915 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |