Learn More
Glucocorticoid Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1512
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-512 detects glucocorticoid receptor (GR) from human, mouse and rat tissues. PA1-512 has been successfully used in Western blot, immunohistochemistry, immunocytochemistry, immunoprecipitation, immunofluorescence, ChIP and gel shift procedures. By Western blot, this antibody detects an ∽97 kDa protein representing GR as well as two other unidentified proteins at ∽135 to ∽145 kDa from rat liver extract. These cross reacting proteins are not believed to be GR or GR precursors as they do not bind [3H]dexamethasone mesylate. Immunohistochemical staining of GR in rat hippocampus using PA1-512 results in staining primarily in the nucleus. PA1-512 does not disrupt receptor-DNA interactions. The PA1-512 immunogen is a synthetic peptide corresponding to residues C(245) K P L I L P D T K P K I K D(259) of human GR. The PA1-512 immunizing peptide (Cat. # PEP-002) is available for use in for neutralization and control experiments. Reconstitute in 50 μL PBS to create a stock of 1 mg/mL.
Glucocorticoid Receptor Polyclonal antibody specifically detects Glucocorticoid Receptor in Human, Mouse, Rat samples. It is validated for ChIP Assay, Gel Shift, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
Glucocorticoid Receptor | |
Polyclonal | |
Unconjugated | |
Nr3c1 | |
cytosolic/nuclear receptor; fb13f09; GCCR; Gcr; GCRST; glucocorticoid nuclear receptor variant 1; glucocorticoid receptor; glucocorticoid receptor 1; glucocorticoid receptor alpha; GR; gr alpha; GR Kit; Grl; Grl1; Grl-1; Nr3c1; Nuclear receptor subfamily 3 group C member 1; nuclear receptor subfamily 3, group C, member 1; nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor); OTTHUMP00000222854; OTTHUMP00000222858; utouto; utut; wu:fb13f09; zgc:113038 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14815, 24413, 2908 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ChIP Assay, Gel Shift, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
1 mg/mL | |
antibody with no preservative | |
P04150, P06536, P06537 | |
Nr3c1 | |
Synthetic peptide corresponding to residues C(245) K P L I L P D T K P K I K D(259) of human GR. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |