Learn More
Glutaminase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544272
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: VNPFPKDRWN NTPMDEALHF GHHDVFKILQ EYQVQYTPQG DSDNGKENQT Sequence homology: Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 93%
Glutaminase Polyclonal antibody specifically detects Glutaminase in Human, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Glutaminase | |
Polyclonal | |
Unconjugated | |
GLS | |
6330442B14; AAD20; AI314027; B230365M23Rik; DKFZp686O15119; FLJ10358; GA; GAC; GAM; GLS; GLS1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; PAG; phosphate-activated glutaminase; RATGLUT | |
Rabbit | |
Affinity Chromatography | |
RUO | |
24398, 2744 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O94925, P13264 | |
GLS | |
Synthetic peptide directed towards the C-terminal of human GLS | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |