Learn More
GFAP Monoclonal Antibody (ASTRO6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512023
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-12023 targets Glial Fibrillary Acidic Protein in IF, IHC (P), and WB applications and shows reactivity with Chicken, Human, Porcine, and Rat samples. The MA5-12023 immunogen is glial Fibrillary Acidic Protein. MA5-12023 was successfully used to detect astrocytes in E18 Sparague Dawley primary cortical cells.
GFAP Monoclonal antibody specifically detects GFAP in Chicken, Human, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
GFAP | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P14136, P47819 | |
GFAP | |
Glial Fibrillary Acidic Protein | |
500 μL | |
Primary | |
Chicken, Human, Porcine, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
ASTRO6 | |
Unconjugated | |
GFAP | |
6330404F12Rik; 65 kDa glutamic acid decarboxylase; AI836096; ALXDRD; Astrocyte or Intermediate Filament Protein; cb345; class III intermediate filament protein; etID36982.3; FLJ45472; GAD(65); Gad2; Gad-2; GAD65; GAD-65; GFAP; GFAP epsilon; gfap protein; gfapl; glial fibrillary acidic protein; Glial Fibrillary Acidic Protein (GFAP); glial fibrillary acidic protein alpha; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; glutamic acid decarboxylase 2; I79_011607; intermediate filament; intermediate filament protein; intermediate filament protein; GFAP; Unknown (protein for MGC:139638); wu:fb34h11; wu:fk42c12; xx:af506734; zgc:110485; zGFAP; intermediate filament protein; zrf-1 antigen | |
Mouse | |
Protein G | |
RUO | |
24387, 2670, 396562, 419969 | |
4° C | |
Liquid |