Learn More
GLI3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528029
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat, mouse ESC, PC-12, Rat2. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (87%), Chicken (91%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GLI3 Polyclonal antibody specifically detects GLI3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GLI3 | |
Polyclonal | |
Unconjugated | |
GLI3 | |
ACLS; add; AI854843; anterior digit pattern deformity; AU023367; Bph; brachyphalangy; extra toes; GCPS; GLI family zinc finger 3; GLI family zinc finger 3 L homeolog; gli3; GLI3 C-terminally truncated form; GLI3 form of 190 kDa; GLI3 form of 83 kDa; GLI3 full length protein; GLI3 full-length protein; gli3.L; GLI3-190; GLI3-83; GLI3FL; GLI-Kruppel family member 3; GLI-Kruppel family member GLI3; GLI-Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome); glioma-associated oncogene family zinc finger 3; im:7142603; neural specific DNA binding protein; neural-specific DNA-binding protein xGLI3; oncogene GLI3; PAPA; PAP-A; PAPA1; PAPB; Pdn; PHS; polydactyly Nagoya; PPDIV; Transcriptional activator GLI3; Transcriptional repressor GLI3R; XELAEV_18031191mg; xgli3; xGLI-3; Xt; Zinc finger protein GLI3; zinc finger protein GLI3; transcriptional activator GLI3; zinc finger transcription factor Gli3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140588, 14634, 2737 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P10071, Q61602 | |
GLI3 | |
Recombinant fragment corresponding to a region within amino acids 243 and 773 of Human GLI3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |