Learn More
GFR alpha-4/GDNF R alpha-4 Mouse anti-Human, Clone: 215725, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1439
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438645
  • £376.32 / 500µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
GFR alpha-4/GDNF R alpha-4 Monoclonal antibody MAB1439 Specifically detects GFR alpha-4/GDNF R alpha-4 in Human samples. It is validated for Western Blotting
Specifications
Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor
215725
Western Blot 1 ug/mL
NP_071422
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human GFR alpha-4/GDNF R alpha-4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) GFR alpha-1, recombinant rat GFR alpha-1, rhGFR alpha-2, rhGFR alpha-3, or recombinant mouse GFR alpha-4 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
GFRA4
Mouse myeloma cell line NS0-derived recombinant human GFR alpha-4/GDNF R alpha-4 Asn24-Ser245 (Ser140Cys)(predicted) Accession # NP_071422
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
64096
Reconstitute at 0.5 mg/mL in sterile PBS.