Learn More
GFR alpha-2/GDNF R alpha-2 Mouse anti-Human, Clone: 129030, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB613
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13441236
  • £337.93 / 500µg
Estimated Shipment
09-07-2019
Description
Description
GFR alpha-2/GDNF R alpha-2 Monoclonal antibody MAB613 Specifically detects GFR alpha-2/GDNF R alpha-2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
GFR alpha-2/GDNF R alpha-2
129030
Western Blot 1 ug/mL
O00451
GDNF family receptor alpha 2, GDNF family receptor alpha-2, GDNF R alpha-2, GDNF receptor alpha-2, GDNF receptor beta, GDNFR-alpha-2, GDNFR-beta, GDNFRBRET ligand 2, GFR alpha2, GFRa-2, GFRA2, GFR-alpha 2, GFR-alpha-2, glial cell line derived neurotrophic factor receptor, beta, Neurturin receptor alpha, NRTNR-alpha, NTNRA, NTNR-alpha, PI-linked cell-surface accessory protein, RETL2TGF-beta-related neurotrophic factor receptor 2, TGF-beta related neurotrophic factor receptor 2, TRN receptor, GPI-anchored, TRNR2NRTNR-ALPHA
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human GFR alpha-2/GDNF R alpha-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) GDNF R alpha , rhGFR alpha-3, rhGFRa-4, and recombinant mouse GFR alpha-2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
GFRA2
Mouse myeloma cell line NS0-derived recombinant human GFR alpha-2/GDNF R alpha-2 Ser22-Ser441 Accession # O00451
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2675
Reconstitute at 0.5 mg/mL in sterile PBS.