Learn More
GFR alpha-1/GDNF R alpha-1 Mouse anti-Human, Clone: 260714, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB7141
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13441676
  • £249.49 / 100µg
Estimated Shipment
27-06-2019
Description
Description
GFR alpha-1/GDNF R alpha-1 Monoclonal antibody MAB7141 Specifically detects GFR alpha-1/GDNF R alpha-1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
GFR alpha-1/GDNF R alpha-1
260714
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
GFRA1
Mouse myeloma cell line NS0-derived recombinant human GFR alpha-1 Asp25-Lys434 Accession # P56159
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2674
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P56159
DKFZp313E1012, DKFZp686J0156, GDNF family receptor alpha 1, GDNF family receptor alpha-1, GDNF R alpha-1, GDNF receptor alpha-1, GDNFRAGFR-ALPHA-1, GDNFR-alpha-1, GDNFRMGC23045, GFR alpha1, GFRa-1, GFRA1, GFR-alpha-1, Glial cell line-derived neurotrophic factor receptor alpha, GPI-linked anchor protein, PI-linked cell-surface accessory protein, RET ligand 1, RET1L, RETL1FLJ10561, TGF-beta related neurotrophic factor receptor 1, TGF-beta-related neurotrophic factor receptor 1, TRNR1FLJ31546
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human GFR alpha-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human GFR alpha-2, -3, -4, or recombinant rat GFR alpha-1 is observed.