Learn More
GFP uv Mouse anti-Multi-species, Clone: 454518, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4240
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430476
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
GFP uv Monoclonal antibody MAB4240 Specifically detects GFP uv in Many Species samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
GFP
454518
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
GFP
E. coli-derived recombinant GFPuv Ala2-Tyr238 Accession # P42212
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects GFPuv in direct ELISAs and Western blots.
Multi-Species
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P42212
eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp)
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Reconstitute at 0.5 mg/mL in sterile PBS.