missing translation for 'onlineSavingsMsg'
Learn More

GFP Monoclonal Antibody (GF28R), HRP, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA515256HRP

Additional Details : Weight : 0.01000kg

Product Code. 15184479

  • £230.00 / 50µg
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MA5-15256-HRP has been successfully used in Western blotting applications with GFP tagged samples.

GFP Monoclonal antibody specifically detects GFP in Tag samples. It is validated for Western Blot.
Specifications

Specifications

GFP
Monoclonal
1 mg/mL
antibody with 0.07% Kathon
Mouse
Protein A
RUO
4° C
Liquid
Tag
Western Blot
GF28R
HRP
GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP
GFP from the jellyfish Aequorea Victoria N-terminal peptide-KLH conjugated.
50 μL
Primary
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers