Learn More
GDF-3 Propeptide Rat anti-Mouse, Clone: 356613, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3719
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430226
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
GDF-3 Propeptide Monoclonal antibody MAB3719 Specifically detects GDF-3 Propeptide in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
GDF-3
356613
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
GDF3
Mouse myeloma cell line NS0-derived recombinant mouse GDF-3 Propeptide Ser23-Lys250 Accession # Q3ZB19
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9573
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q3ZB19
GDF3, GDF-3, growth differentiation factor 3, growth/differentiation factor 3, KFS3, MCOP7, MCOPCB6, Vgr-2
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse GDF-3 Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) GDF-1, rmGDF-1pro, rmGDF-3, rmGDF-5, rmGDF-6, rmGDF-7, rmGDF-8, or rmGDF-9 is observed.