Learn More
GATA-1 Rat anti-Human, Clone: 234732, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1779
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458915
  • £249.49 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
GATA-1 Monoclonal antibody MAB1779 Specifically detects GATA-1 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
GATA-1
234732
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
GATA1
E. coli-derived recombinant human GATA-1 Met1-Ser413 Accession # P15976
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2623
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunocytochemistry
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
P15976
Eryf1, ERYF1GATA-binding protein 1 (globin transcription factor 1), GATA binding protein 1 (globin transcription factor 1), GATA1, GATA-1erythroid transcription factor, GATA-binding factor 1, GF-1, GF1globin transcription factor 1, NF-E1 DNA-binding protein, NFE1erythroid transcription factor 1, transcription factor GATA1, XLTT
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human GATA-1 in direct ELISAs. In Western blots, this antibody does not cross-react with rhGATA-2 or rhGATA-6.