missing translation for 'onlineSavingsMsg'
Learn More

GASP-2/WFIKKN Mouse anti-Human, Clone: 289827, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB21361

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459205

  • £337.93 / 500µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

GASP-2/WFIKKN Monoclonal antibody MAB21361 Specifically detects GASP-2/WFIKKN in Human samples. It is validated for ELISA Capture
Specifications

Specifications

GASP-2/WFIKKN
289827
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
WFIKKN1
Mouse myeloma cell line NS0-derived recombinant human GASP-2/WFIKKN Ala20-Asp548 Accession # Q96NZ8
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
117166
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q96NZ8
C16orf12, Chromosome 16 Open Reading Frame 12, GASP2, GASP-2, Growth And Differentiation Factor-Associated Serum Protein 2, hGASP-2, RJD2, WAP Four-Disulfide Core Domain 20A, WAP, Follistatin/Kazal, Immunoglobulin, Kunitz And Netrin Domain Containing 1, WAP, FS, Ig, KU, And NTR-Containing Protein, WFDC20A, WFIKKN, WFIKKN1
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human GASP-2/WFIKKN in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human GASP-1 is observed.
Human
Videos

For Research Use Only