missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH Loading Control Monoclonal Antibody (GA1R), DyLight™ 488, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515738D488
156.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-15738-D488 has successfully been used for immunofluorescence. DyLight 488 has an excitation/emission of 493/518nm.
GAPDH Loading Control Monoclonal antibody specifically detects GAPDH Loading Control in Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast samples. It is validated for Immunocytochemistry.Specifications
GAPDH Loading Control | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
O14556, P00356, P04406, P04797, P16858, P17244, P46406, Q64467, Q9ESV6 | |
GAPDH, GAPDHS | |
Recombinant GAPDH | |
50 μL | |
Primary | |
Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast | |
Antibody | |
IgG1 |
Immunocytochemistry | |
GA1R | |
DyLight 488 | |
GAPDHS | |
38 kDa BFA-dependent ADP-ribosylation substrate; aging-associated gene 9 protein; BARS-38; bb02e05; cb350; cb609; CDABP0047; EC 1.2.1.12; epididymis secretory protein Li 278; epididymis secretory sperm binding protein Li 162eP; fb71f08; fk58c09; G3PD; G3PDH; GAPD; GAPD2; gapdh; GAPDH2; GAPDH-2; GAPDHS; Gapds; Gapd-s; glceraldehyde-3-phosphate dehydrogenase; glyceraldehyde 3-phosphate dehydrogenase; glyceraldehyde 3-phosphate dehydrogenase, testis-specific; glyceraldehyde phosphate dehydrogenase; glyceraldehyde-3-phosphate dehydrogenase; glyceraldehyde-3-phosphate dehydrogenase (G3PDH); glyceraldehyde-3-phosphate dehydrogenase 2; glyceraldehyde-3-phosphate dehydrogenase GAPDH; glyceraldehyde-3-phosphate dehydrogenase like-17 protein; glyceraldehyde-3-phosphate dehydrogenase type 2; glyceraldehyde-3-phosphate dehydrogenase, spermatogenic; glyceraldehyde-3-phosphate dehydrogenase, testis-specific; glyceraldehyde-phosphate-dehydrogenase; glycerine aldehyde 3-phosphate dehydrogenase; HEL-S- | |
Mouse | |
Protein A | |
RUO | |
100009074, 100352213, 100736557, 100767862, 14433, 14447, 24383, 2597, 26330, 374193, 66020 | |
4° C, do not freeze | |
Liquid |