missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALK2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513845
Code : LE
Additional Details : Weight : 0.01000kg
Description
GALK2 is a highly efficient N-acetylgalactosamine (GalNAc) kinase, which has galactokinase activity when galactose is present at high concentrations.Specifications
GALK2 | |
Polyclonal | |
Unconjugated | |
Q01415 | |
GALK2 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human GALK2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
2 mg/mL | |
GALK2 | |
N-acetylgalactosamine kinase; GalNAc kinase; Galactokinase 2; GK2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2585 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |