missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA515321
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat based on sequence homology.
GAK Polyclonal antibody specifically detects GAK in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
GAK | |
Polyclonal | |
Unconjugated | |
GAK | |
auxilin-2; cyclin G associated kinase; cyclin G-associated kinase; cyclin-G-associated kinase; D130045N16Rik; DNAJ26; DNAJC26; GAK | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2580 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
O14976 | |
GAK | |
KLH conjugated synthetic peptide between 1140-1170 amino acids from the C-terminal region of human GAK | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |