missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GADD34 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530486
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GADD34 Polyclonal antibody specifically detects GADD34 in Human samples. It is validated for Western Blot.Specifications
GADD34 | |
Polyclonal | |
Unconjugated | |
PPP1R15A | |
9630030H21; Gadd34; growth arrest and DNA damage-inducible protein GADD34; growth arrest and DNA-damage-inducible 34; Myd116; myeloid differentiation primary response gene 116; myeloid differentiation primary response protein MyD116; Myeloid differentiation primary response protein MyD116 homolog; Peg3; PEG-3; PPP1R15A; PR15A; Progression elevated gene 3 protein; protein phosphatase 1 regulatory subunit 15A; protein phosphatase 1, regulatory (inhibitor) subunit 15A; protein phosphatase 1, regulatory subunit 15A; RGD1624209 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O75807 | |
PPP1R15A | |
Recombinant fragment corresponding to a region within amino acids 95 and 312 of Human GADD34 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |