missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FZD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533555
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Rat, Dolphin, Hamster, Elephant, Panda, Dog, Bovine (100%).
FZD4 Polyclonal antibody specifically detects FZD4 in Bovine, Canine, Hamster, Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
FZD4 | |
Polyclonal | |
Unconjugated | |
FZD4 | |
CD344; CD344 antigen; EVR1; FEVR; frizzled 4, seven transmembrane spanning receptor; frizzled class receptor 4; frizzled family receptor 4; frizzled homolog 4; frizzled homolog 4 (Drosophila); frizzled receptor 4; frizzled-4; Fz4; Fz-4; Fzd4; FZD4S; FzE4; GPCR; hFz4; mFz4; MGC34390; rFz4; WNT receptor frizzled-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100756296, 445416, 485149, 8322 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 0.1% sodium azide | |
Q9ULV1 | |
FZD4 | |
Synthetic 18 amino acid peptide from N-terminal extracellular domain of human FZD4 / Frizzled 4. | |
50 μg | |
Primary | |
Bovine, Canine, Hamster, Human, Non-human Primate | |
Antibody | |
IgG |