Learn More
FXR2 Monoclonal Antibody (A42), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA15773
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-5773 detects FXR2 in human samples and is predicted to function in mouse samples. MA1-5773 has been successfully used in Western blot, immunoprecipitation, immunofluorescence and ELISA procedures. MA1-5773 detects a band of approximately 92 kDa. The MA1-5773 immunogen is His6-FXR2s fusion protein produced in a bacterial pET system.
FXR2 Monoclonal antibody specifically detects FXR2 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
FXR2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P51116 | |
FXR2 | |
His6-FXR2s fusion protein produced in a bacterial pET system with SP2/0 cells | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
A42 | |
Unconjugated | |
FXR2 | |
FMR1 autosomal homolog 2; FMR1L2; fragile X mental retardation gene 2, autosomal homolog; fragile X mental retardation syndrome-related protein 2; fragile X mental retardation, autosomal homolog 2; fragile X mental retardation-related protein 2; fragile X-mental retardation 1-like 2; Fxr2; Fxr2h; FXR2P | |
Mouse | |
Protein A | |
RUO | |
9513 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |