missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513410
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
FXN Polyclonal antibody specifically detects FXN in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
FXN | |
Polyclonal | |
Unconjugated | |
FXN | |
CyaY; d-FXN; DKEYP-118A9.1; FA; FARR; frataxin; Frataxin intermediate form; Frataxin mature form; Frataxin(56-210); Frataxin(78-210); Frataxin(81-210); Frataxin, mitochondrial; FRDA; Friedreich ataxia; Friedreich ataxia protein; FXN; i-FXN; m56-FXN; m78-FXN; m81-FXN; RGD1565754; X25; zgc:158218 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2395 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q16595 | |
FXN | |
KLH conjugated synthetic peptide between 143-172 amino acids from the C-terminal region of human FXN | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |