missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513515
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rabbit based on sequence homology.
FUT1 Polyclonal antibody specifically detects FUT1 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
FUT1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P19526 | |
FUT1 | |
KLH conjugated synthetic peptide between 164-195 amino acids from the central region of human FUT1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
FUT1 | |
2-alpha-L-fucosyltransferase; alpha (1,2) fucosyltransferase; alpha 1,2-fucosyltransferase A; alpha 12-fucosyltransferase; alpha(1,2) fucosyltransferase 1; alpha(1,2)FT 1; alpha12-fucosyltransferase a; beta-galactoside alpha-1,2-fucosyltransferase; blood group H alpha 2-fucosyltransferase; Fta; Fucosyltransferase 1; fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase); fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group); fucosyltransferase 1 (H blood group); FUT1; Futa; galactoside 2-alpha-L-fucosyltransferase 1; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1; H; H transferase; HH; HSC | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2523 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |