Learn More
Fumarase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522091
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, mosue liver, rat liver. Predicted reactivity: Rat (96%), Zebrafish (86%), Pig (97%), Chicken (89%), Sheep (97%), Chimpanzee (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Fumarase Polyclonal antibody specifically detects Fumarase in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Fumarase | |
Polyclonal | |
Unconjugated | |
FH | |
EF-3; Fh; Fh1; Fh-1; FMRD; fumarase; fumarate hydratase; fumarate hydratase 1; fumarate hydratase, mitochondrial; HLRCC; LRCC; MCL; MCUL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14193, 14194, 2271, 24368 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P07954, P14408, P97807 | |
FH, Fh1 | |
Recombinant fragment corresponding to a region within amino acids 89 and 365 of Fumarate hydratase (Uniprot ID#P07954) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |