missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Light Chain Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519059
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
Ferritin Light Chain Polyclonal antibody specifically detects Ferritin Light Chain in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Ferritin Light Chain | |
Polyclonal | |
Unconjugated | |
FTL | |
ferritin L subunit; Ferritin L subunit 1; ferritin L-chain; ferritin light chain; Ferritin light chain 1; ferritin light polypeptide-like 3; ferritin, light polypeptide; fril; Ftl; Ftl1; Ftl-1; L-ferritin; LFTD; NBIA3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14325, 2512, 29292 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P02792, P02793, P29391 | |
FTL, Ftl1 | |
Synthetic peptide sequence (GEYLFERLTLKHD) corresponding to the C-terminus amino acids of FTL | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |